missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Malonyl dichloride, 97%, Thermo Scientific Chemicals

$229.68 - $751.52

Chemical Identifiers

CAS 1663-67-8
Molecular Formula C3H2Cl2O2
Molecular Weight (g/mol) 140.95
MDL Number MFCD00000735
InChI Key SXYFKXOFMCIXQW-UHFFFAOYSA-N
Synonym malonyl chloride, malonyl dichloride, malonoyl chloride, malonic acid dichloride, malonoyl dichloride, malonic acid chloride, malonylchloride, methane-1,1-dicarbonyl chloride, malonylchlorid, malonyldichloride
PubChem CID 74269
IUPAC Name propanedioyl dichloride
SMILES C(C(=O)Cl)C(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC125280250
SDS View Documents
Thermo Scientific Chemicals
125280250
25 g Glass bottle
Each for $229.68
 
AC125281000
SDS View Documents
Thermo Scientific Chemicals
125281000
100 g Glass bottle
Each for $751.52
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1663-67-8
140.95
SXYFKXOFMCIXQW-UHFFFAOYSA-N
74269
C(C(=O)Cl)C(=O)Cl
C3H2Cl2O2
MFCD00000735
malonyl chloride, malonyl dichloride, malonoyl chloride, malonic acid dichloride, malonoyl dichloride, malonic acid chloride, malonylchloride, methane-1,1-dicarbonyl chloride, malonylchlorid, malonyldichloride
propanedioyl dichloride

Specifications

1663-67-8
1.4500g/mL
47°C
96% min. (Argentometry)
C3H2Cl2O2
ClCOCH2COCl
25 g
12,291
malonyl chloride, malonyl dichloride, malonoyl chloride, malonic acid dichloride, malonoyl dichloride, malonic acid chloride, malonylchloride, methane-1,1-dicarbonyl chloride, malonylchlorid, malonyldichloride
SXYFKXOFMCIXQW-UHFFFAOYSA-N
propanedioyl dichloride
74269
97%
Malonyl dichloride
Brown to Yellow
47.0°C (15.0 mmHg)
Authentic
Glass bottle
1.4620 to 1.4660
MFCD00000735
02, I, 252
1.45
Solubility in water: reacts violently. Other solubilities: soluble in ether
C(C(=O)Cl)C(=O)Cl
140.95
140.95
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 216-772-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only