missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganese(II) bromide, 99%, anhydrous, Thermo Scientific Chemicals

$153.78 - $153.78

Chemical Identifiers

CAS 13446-03-2
Molecular Formula Br2Mn
Molecular Weight (g/mol) 214.76
MDL Number MFCD00011113
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC207860250
SDS View Documents
Thermo Scientific Chemicals
207860250
25 g Glass bottle
Each for $153.78
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13446-03-2
Br2Mn
214.76
MFCD00011113

Specifications

Pink
Powder
99%
MnBr2
15, 5790
214.76
99%
4.38
Manganese(II) bromide, Anhydrous
13446-03-2
25 g
Br2Mn
MFCD00011113
Solubility in water: soluble
214.76
Glass bottle
4.3800g/mL
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS Signal Word: Warning

EINECSNumber : 236-591-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only