missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganese(II) bromide hydrate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04024522

 View more versions of this product

Catalog No. AA4024522


Explore available promotions

Description

Description

It is an important raw material and intermediate used in organic synthesis. It can be used in place of palladium in the stille reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Manganese(II) bromide hydrate
10031-20-6
Crystalline Aggregates
Br2H8MnO4
14,5725
Soluble in water, alcohol
O.O.O.O.[Mn++].[Br-].[Br-]
286.81
98%
Hygroscopic
64.3°C (decomposition)
Pink
100 g
MFCD00149791
manganese bromide tetrahydrate, unii-8308v6rbj5, manganese ii bromide hexahydrate, manganous bromide tetrahydrate, manganese dibromide tetrahydrate, manganese bromide mnbr2 , tetrahydrate, manganese 2+ ion tetrahydrate dibromide
HHDPJRSXPOGIOP-UHFFFAOYSA-L
manganese(2+) tetrahydrate dibromide
214.76 (Anhydrous)
Odorless

Chemical Identifiers

10031-20-6
286.81
HHDPJRSXPOGIOP-UHFFFAOYSA-L
manganese(2+) tetrahydrate dibromide
Br2H8MnO4
MFCD00149791
manganese bromide tetrahydrate, unii-8308v6rbj5, manganese ii bromide hexahydrate, manganous bromide tetrahydrate, manganese dibromide tetrahydrate, manganese bromide mnbr2 , tetrahydrate, manganese 2+ ion tetrahydrate dibromide
O.O.O.O.[Mn++].[Br-].[Br-]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

EINECSNumber : 236-591-9

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only