missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganese(II) bromide hydrate, 98%, Thermo Scientific Chemicals

$75.42 - $679.04

Chemical Identifiers

CAS 10031-20-6
Molecular Formula Br2H8MnO4
Molecular Weight (g/mol) 286.81
MDL Number MFCD00149791
InChI Key HHDPJRSXPOGIOP-UHFFFAOYSA-L
Synonym manganese bromide tetrahydrate, unii-8308v6rbj5, manganese ii bromide hexahydrate, manganous bromide tetrahydrate, manganese dibromide tetrahydrate, manganese bromide mnbr2 , tetrahydrate, manganese 2+ ion tetrahydrate dibromide
IUPAC Name manganese(2+) tetrahydrate dibromide
SMILES O.O.O.O.[Mn++].[Br-].[Br-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4024514
View Documents
Thermo Scientific Chemicals
04024514
25 g
Each for $75.42
 
AA4024522
View Documents
Thermo Scientific Chemicals
04024522
100 g
Each for $200.81
 
AA4024536
View Documents
Thermo Scientific Chemicals
04024536
500 g
Each for $679.04
 
Description

Description

It is an important raw material and intermediate used in organic synthesis. It can be used in place of palladium in the stille reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10031-20-6
286.81
HHDPJRSXPOGIOP-UHFFFAOYSA-L
manganese(2+) tetrahydrate dibromide
Br2H8MnO4
MFCD00149791
manganese bromide tetrahydrate, unii-8308v6rbj5, manganese ii bromide hexahydrate, manganous bromide tetrahydrate, manganese dibromide tetrahydrate, manganese bromide mnbr2 , tetrahydrate, manganese 2+ ion tetrahydrate dibromide
O.O.O.O.[Mn++].[Br-].[Br-]

Specifications

64.3°C (decomposition)
Pink
25 g
MFCD00149791
manganese bromide tetrahydrate, unii-8308v6rbj5, manganese ii bromide hexahydrate, manganous bromide tetrahydrate, manganese dibromide tetrahydrate, manganese bromide mnbr2 , tetrahydrate, manganese 2+ ion tetrahydrate dibromide
HHDPJRSXPOGIOP-UHFFFAOYSA-L
manganese(2+) tetrahydrate dibromide
214.76 (Anhydrous)
Odorless
Manganese(II) bromide hydrate
10031-20-6
Crystalline Aggregates
Br2H8MnO4
14,5725
Soluble in water, alcohol
O.O.O.O.[Mn++].[Br-].[Br-]
286.81
98%
Hygroscopic
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

EINECSNumber : 236-591-9

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only