Learn More
Manganese(IV) oxide, 97%, Thermo Scientific Chemicals
$133.32 - $958.74
Chemical Identifiers
CAS | 1313-13-9 |
---|---|
Molecular Formula | MnO2 |
Molecular Weight (g/mol) | 86.94 |
MDL Number | MFCD00003463 |
InChI Key | NUJOXMJBOLGQSY-UHFFFAOYSA-N |
Synonym | manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein |
PubChem CID | 14801 |
IUPAC Name | dioxomanganese |
SMILES | O=[Mn]=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1076522
|
Thermo Scientific Chemicals
A1076522 |
100 g |
Each for $133.32
|
|
||||||
AAA1076536
|
Thermo Scientific Chemicals
A1076536 |
500 g |
Each for $248.36
|
|
||||||
AAA107650E
|
Thermo Scientific Chemicals
A107650E |
2500 g |
Each for $343.32
|
|
||||||
AAA10765A9
|
Thermo Scientific Chemicals
A10765A9 |
10 kg |
Each for $958.74
|
|
||||||
Description
Manganese(IV) oxide is used as a pigment and as a reagent inorganic synthesis especially for the oxidation of allylic alcohols. It is also used for dry -cell batteries such as the alkaline battery and the zinc-carbon battery. It is involved in the preparation of other manganese compounds. It is a precursor to ferromanganese and related alloys, which is useful in the steel industry. It catalyses the decomposition of hydrogen peroxide into oxygen and water. Further, it is used to prepare manganese(II) sulfate, manganese(II) chloride and potassium permanganate. In addition to this, it is used in ceramics and glass manufacturing as an inorganic pigment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsManganese(IV) oxide is used as a pigment and as a reagent inorganic synthesis especially for the oxidation of allylic alcohols. It is also used for dry -cell batteries such as the alkaline battery and the zinc-carbon battery. It is involved in the preparation of other manganese compounds. It is a precursor to ferromanganese and related alloys, which is useful in the steel industry. It catalyses the decomposition of hydrogen peroxide into oxygen and water. Further, it is used to prepare manganese(II) sulfate, manganese(II) chloride and potassium permanganate. In addition to this, it is used in ceramics and glass manufacturing as an inorganic pigment.
Solubility
Insoluble in water.
Notes
Incompatible with strong acids, strong reducing agents and organic materials.
Chemical Identifiers
1313-13-9 | |
86.94 | |
NUJOXMJBOLGQSY-UHFFFAOYSA-N | |
14801 | |
O=[Mn]=O |
MnO2 | |
MFCD00003463 | |
manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein | |
dioxomanganese |
Specifications
535°C (decomposition) | |
100 g | |
MFCD00003463 | |
manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein | |
NUJOXMJBOLGQSY-UHFFFAOYSA-N | |
dioxomanganese | |
14801 | |
98% | |
Manganese (IV) oxide |
1313-13-9 | |
MnO2 | |
14,5730 | |
Insoluble in water. | |
O=[Mn]=O | |
86.94 | |
86.94 | |
5.026 |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P260-P264b-P270-P271-P301+P312-P304+P340-P312-P314-P330-P501c
H302+H332-H373
EINECSNumber : 215-202-6
RTECSNumber : OP0350000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only