missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Manganese powder, -325 mesh, 99.95% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03631522
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Manganese | |
7.3g/cm3 at 20°C | |
Mn | |
100 g | |
PWHULOQIROXLJO-UHFFFAOYSA-N | |
manganese | |
23930 | |
-325 Mesh |
7439-96-5 | |
99.95%, metal basis | |
MFCD00011111 | |
colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds | |
[Mn] | |
54.94 | |
CHEBI:35154 | |
Powder |
Chemical Identifiers
7439-96-5 | |
54.94 | |
PWHULOQIROXLJO-UHFFFAOYSA-N | |
23930 | |
manganese |
Mn | |
MFCD00011111 | |
colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds | |
CHEBI:35154 | |
[Mn] |
Safety and Handling
Take precautionary measures against static discharges.
P210-P240-P241-P264b-P280-P305+P351+P338-P370+P378q
H228-H319
DOTInformation : DOT Class: 4.1; Packaging Group: III
EINECSNumber : 231-105-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only