missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Neutec Group Masticator Paddle Lab Blenders

Used for solid and semisolid samples by applying pressure to samples within sterile, disposable bags

Supplier:  Neutec Group 9000471

 View more versions of this product

Catalog No. 14-910-426


Explore available promotions

Description

Description

  • Compact size
  • Reduce work load on lab technician
  • Long tradition of reliability and robustness
  • Use metal springs to move the paddles and not a belt
  • Small bench space required due to its long rather than wide design
  • Viewing window option (panoramic models) for better monitoring of sample condition
  • Works with large selection of Lab Blending Bags

Warranty

3 years

Specifications

Specifications

Lab Blender
39.8 x 23.8 x 26 cm (15.7 x 9.4 x 10.2 in.)
Basic
37.5 lb.
1500 rpm
Analog timer
8.5 (50Hz) 10 (60Hz)
17 kg
Videos
SDS
Documents

Documents

Promotions

Promotions