missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Neutec Group Masticator Paddle Lab Blenders

Used for solid and semisolid samples by applying pressure to samples within sterile, disposable bags

$9788.94 - $14088.96

Specifications

Product Type Lab Blender
Dimensions (D x W x H) 39.8 x 23.8 x 26 cm (15.7 x 9.4 x 10.2 in.)
View More Specs

Products 2
Catalog Number Mfr. No. Model No. of Strokes Speed Display Price Quantity  
Catalog Number Mfr. No. Model No. of Strokes Speed Display Price Quantity  
14-910-429
View Documents
Neutec Group
9000522
Silver Panoramic 5/6/7/8 500 to 1500 rpm Digital N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14-910-426
View Documents
Neutec Group
9000471
Basic 8.5 (50Hz) 10 (60Hz) 1500 rpm Analog timer
Each for $9,788.94
 
Description

Description

  • Compact size
  • Reduce work load on lab technician
  • Long tradition of reliability and robustness
  • Use metal springs to move the paddles and not a belt
  • Small bench space required due to its long rather than wide design
  • Viewing window option (panoramic models) for better monitoring of sample condition
  • Works with large selection of Lab Blending Bags
Specifications

Specifications

Lab Blender
39.8 x 23.8 x 26 cm (15.7 x 9.4 x 10.2 in.)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'