missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Matrix™ 2D Barcoded Clear Polypropylene Open-Top Storage Tubes with Installed DuraSeal Septum Seal

Store and track sample volumes with these 2D Barcoded Clear Polypropylene Storage Tubes.

Supplier:  Thermo Scientific™ 3737

 View more versions of this product

Catalog No. 14754549


Explore available promotions

Description

Description

Store, track and access samples with ease with Thermo ScientificTM MatrixTM 2D Barcoded Clear Open-Top Storage Tubes with DuraSeal Septum installed. The permanently-installed DuraSeal septum allows easy access to tube contents and enables long-term storage at temperatures down to -150°C. The proprietary barcoding process produces a permanent, high-contrast 2D barcode offering maximum reliability and traceability of compound, biological and genomic samples.

Tracking:
  • A permanently-bonded, unique 2D barcode is laser-etched onto the base of every Matrix 2D barcoded storage tube to securely identify and track samples
  • Corresponding with a laboratory's throughput needs, Thermo Scientific™ VisionMate™ 2D barcode readers instantly decode each tube's 2D barcode
  • A range of VisionMate 2D barcode readers is available to meet your application and throughput needs.
  • Scan the tube into any application or database, or integrate via various methods
  • VisionMate systems are flexible enough to allow 2D barcode data to be integrated into various applications, databases and automated systems
  • Once sample data is stored, user can scan the tube with the database
  • Tracking system includes a "find" feature to instantly access sample information and identify the contents of each storage tube
Superior Storage Rack Design:
  • Matrix 96-format 2D barcoded storage tubes are available in a proprietary specially-designed, stackable, microplate footprint
  • Latch racks save precious space in your freezer, refrigerator, compound store or on the bench top
  • The latch rack design enables manual multichannel pipette access to 2D tubes and eliminates contamination risk with lid design that does not contact the benchtop
  • The latch rack lid can be lifted robotically for 2D tube access with automated liquid handling systems and high throughput applications
Stringent Quality Control:
  • Every 2D barcoded storage tube is scanned to guarantee readability
  • Each code is checked against a full database of all previously assigned 2D codes to ensure zero duplicates across the entire line of Matrix 2D barcoded storage tubes
Storage Tracking Solutions for any Laboratory:
  • Custom 2D coding options are available to meet your laboratory's individual database or tracking requirements
  • 2D storage tubes are compatible with Thermo Scientific freezer racks that allow optimal utility of storage space
Specifications

Specifications

500 μL
10 latch racks of 96 tubes/cs.
2D BarCoded
Sterile
Polypropylene
w/DuraSeal™ Septum Inserted
Polypropylene
V-bottom
Matrix™
5 Case
16907 fl oz.
Videos
SDS
Documents

Documents

Promotions

Promotions