Thermo Scientific™ Matrix™ Reagent Reservoirs
Accommodate a variety of applications with these uniquely designed, innovative reagent reservoirs.
Supplier:  Thermo Scientific™ 809611
 View more versions of this product
Catalog No. 14-387-072
Explore available promotions
Description
Description
Accomodate a variety of applications with uniquely designed, innovative Thermo Scientific™ Matrix Reagent Reservoirs. These sterile reservoirs offer an extra-wide base to add rigidity and stability, helping to avoid spills. The 25mL divided reservoir allows the pipetting of two different reagents with up to eight channels on one side, and up to four on the other; maximizing recovery if using only a single channel and a small amount of reagent.
  • Trough within a trough maximizes amount of liquid accessible to pipet tips when using small amounts of reagent
  • Pour-off spouts on all four corners reduce spillage when pouring reagents out of reservoir
  • Graduations on inside wall enable quick measurement of remaining liquid
  • Sterile reservoirs are packaged in snap and tear bags, maintaining sterility while being easy to open
Specifications
Specifications
25 mL
Sterile
50 Individually wrapped
Polystyrene
Reagent Reservoir with Divider