Thermo Scientific™ Matrix™ Reagent Reservoirs
Accommodate a variety of applications with these uniquely designed, innovative reagent reservoirs.
$244.22 - $637.53
Catalog Number | Mfr. No. | Capacity (Metric) | Material | Type | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Material | Type | Price | Quantity | ||||
14-387-065
|
Thermo Scientific™
8075 |
75 mL | Polypropylene | Reagent Reservoir |
Case of 10 Each for $447.38
|
|
||||
50824108
|
Thermo Scientific™
8076 |
75 mL | Polypropylene | Reservoir Lid |
Case of 10 Each for $244.22
|
|
||||
14-387-067
|
Thermo Scientific™
MX8085 |
100 mL | Polystyrene | Reagent Reservoir |
Case of 100 Each for $400.11
|
|
||||
14-387-068
|
Thermo Scientific™
MX8086 |
100 mL | Polystyrene | Reagent Reservoir |
Case of 50 Each for $596.94
|
|
||||
14-387-069
|
Thermo Scientific™
809311 |
25 mL | Polystyrene | Reagent Reservoir |
Case of 100 Each for $412.30
|
|
||||
14-387-070
|
Thermo Scientific™
8094 |
25 mL | Polystyrene | Reagent Reservoir |
Case of 50 Each for $401.41
|
|
||||
14-387-071
|
Thermo Scientific™
8095 |
25 mL | Polystyrene | Reagent Reservoir with Divider |
Case of 100 Each for $412.30
|
|
||||
14-387-072
|
Thermo Scientific™
809611 |
25 mL | Polystyrene | Reagent Reservoir with Divider |
Case of 50 Each for $637.53
|
|
||||
Description
Accomodate a variety of applications with uniquely designed, innovative Thermo Scientific™ Matrix Reagent Reservoirs. These sterile reservoirs offer an extra-wide base to add rigidity and stability, helping to avoid spills. The 25mL divided reservoir allows the pipetting of two different reagents with up to eight channels on one side, and up to four on the other; maximizing recovery if using only a single channel and a small amount of reagent.
- Trough within a trough maximizes amount of liquid accessible to pipet tips when using small amounts of reagent
- Pour-off spouts on all four corners reduce spillage when pouring reagents out of reservoir
- Graduations on inside wall enable quick measurement of remaining liquid
- Sterile reservoirs are packaged in snap and tear bags, maintaining sterility while being easy to open