Thermo Scientific™ MaxQ™ HP Orbital Shakers Universal Platforms
Mix and match different size labware on a single platform with these orbital shakers universal platforms.
Supplier:  Thermo Scientific™ 238002
 View more versions of this product
Catalog No. 238002
Explore available promotions
Description
Description
Mix and match different size labware on a single platform with flexible Thermo Scientific™ MaxQ™ HP Orbital Shakers Universal Platforms.
Specifications
Specifications
25, 50, 125, 250/300, 500 and 2800 mL; 1, 2, 4, 5, and 6 L; Microplate
Certificate of Analysis (COA), Certificate of Conformance (COC)
36 in.
24 in.
Universal Platform without Clamps, 24 x 36 in.
Universal Platform
24 x 36 in.
91.4 cm
61 cm
MaxQ™ HP Orbital Shakers