Thermo Scientific™ MaxQ™ HP Orbital Shakers Universal Platforms
Mix and match different size labware on a single platform with these orbital shakers universal platforms.
Supplier:  Thermo Scientific™ 238008
 View more versions of this product
Catalog No. 238008
Explore available promotions
Description
Description
Mix and match different size labware on a single platform with flexible Thermo Scientific™ MaxQ™ HP Orbital Shakers Universal Platforms.
Specifications
Specifications
Universal Platform
18 x 24 in.
61 cm
45.7 cm
MaxQ™ HP Orbital Shakers
Certificate of Analysis (COA), Certificate of Conformance (COC)
24 in.
18 in.
Universal Platform without Clamps, 18 x 24 in.