Thermo Scientific™ MaxQ™ Shaker Universal Platforms
Mix and match different size labware on a single platform with flexible Thermo Scientific™ MaxQ™ Shaker Universal Platforms.
Supplier: Thermo Scientific™ 30110
Description
Mix and match different size labware on a single platform with flexible Thermo Scientific™ MaxQ™ Shaker Universal Platforms. With exceptional innovation, reliability and accuracy, these platforms are the optimal choice for a busy lab.
For use with MaxQ Shakers as indicated. Choose the platform that best suits your needs.
Ordering Information:
Purchase clamps separately.
Specifications
Universal Platform | |
18 x 18 in. (45.7 x 45.7 cm) | |
45.7 cm | |
45.7 cm | |
MaxQ™ 2000, 2506, 2508, 4000, 6000 Shaker |
Certificate of Analysis (COA), Certificate of Conformance (COC) | |
18 in. | |
18 in. | |
Universal Platform without Clamps, 18 x 18 in. |