Thermo Scientific™ MaxQ™ Shaker Universal Platforms
Mix and match different size labware on a single platform with flexible Thermo Scientific™ MaxQ™ Shaker Universal Platforms.
Supplier: Thermo Scientific™ 30106
Description
Mix and match different size labware on a single platform with flexible Thermo Scientific™ MaxQ™ Shaker Universal Platforms. With exceptional innovation, reliability and accuracy, these platforms are the optimal choice for a busy lab.
For use with MaxQ Shakers as indicated. Choose the platform that best suits your needs.
Ordering Information:
Purchase clamps separately.
Specifications
Shaker Platform | |
18 x 24 in. | |
45.7 cm | |
60.9 cm | |
MaxQ 2000, 2506, 2508 |
18 x 24 in. (45.7 x 60.9 cm) | |
18 in. | |
24 in. | |
Universal Platform without Clamps, 18 x 24 in. |