Integra™ Miltex™ MeisterHand™ Iris Scissors
Offer outstanding distal cutting capability
Supplier:  Integra™ Miltex™ MH5302
 View more versions of this product
Catalog No. 12-460-656
Explore available promotions
Description
Description
  • Sharp/sharp scissors available in straight and curved
  • Tungsten carbids scissors have tungsten carbide inserts seamlessly bonded to upper and lower blades and feature gold ring handles
  • SuperCut scissors have a razor sharp edge that cuts effortlessly through tissue; lower blade has micro fine serrations to help prevent tissue slippage
  • Stainless steel
  • Non-glaring matte finish
Order Info
Cold or chemical sterilization not recommended for Carb-N-Sert scissors. SuperCut scissiors designed to cut tissue only; not recommended for use on suture material. Dropping or mishandling will cause the razor-shapr edge to nick or become irregular.
Specifications
Specifications
Iris Scissors
4 in.
Stainless Steel
Curved
10.2 cm
Surgical, Ophthalmic
Product Suggestions
Product Suggestions