Integra™ Miltex™ MeisterHand™ Iris Scissors
Offer outstanding distal cutting capability
$137.38 - $147.03
Catalog Number | Mfr. No. | Blade Style | Length (Metric) | Product Type | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Blade Style | Length (Metric) | Product Type | Price | Quantity | ||||
12-460-659
|
Integra™ Miltex™
MH5306 |
Curved | 11.43 cm | Iris Scissors |
Each for $145.38
|
|
||||
12-460-655
|
Integra™ Miltex™
MH5300 |
Straight | 10.2 cm | Iris Scissors |
Each for $137.38
|
|
||||
12-460-656
|
Integra™ Miltex™
MH5302 |
Curved | 10.2 cm | Iris Scissors |
Each for $147.03
|
|
||||
12-460-657
|
Integra™ Miltex™
MH5304 |
Straight | 11.4 cm | Iris Scissors |
Each for $140.44
|
|
||||
Description
- Sharp/sharp scissors available in straight and curved
- Tungsten carbids scissors have tungsten carbide inserts seamlessly bonded to upper and lower blades and feature gold ring handles
- SuperCut scissors have a razor sharp edge that cuts effortlessly through tissue; lower blade has micro fine serrations to help prevent tissue slippage
- Stainless steel
- Non-glaring matte finish
Order Info
Cold or chemical sterilization not recommended for Carb-N-Sert scissors. SuperCut scissiors designed to cut tissue only; not recommended for use on suture material. Dropping or mishandling will cause the razor-shapr edge to nick or become irregular.
Specifications
Stainless Steel |