Integra™ Miltex™ MeisterHand™ Mayo Dissecting Scissors
distal cutting
$124.79 - $337.59
Specifications
For Use With (Application) Dissecting
View More Specs
Products 5
Catalog Number Mfr. No. Length (Metric) Material Blade Style Product Type Price Quantity  
Catalog Number Mfr. No. Length (Metric) Material Blade Style Product Type Price Quantity  
12-460-638
View Documents
Integra™ Miltex™
MH5122
14.6 cm Stainless Steel Curved, Beveled Dissecting Scissors
Each for $139.29
12-460-637
View Documents
Integra™ Miltex™
MH5120
14.6 cm Stainless Steel Straight, Beveled Dissecting Scissors
Each for $124.79
12-460-640
View Documents
Integra™ Miltex™
MH5124
17 cm Stainless Steel Straight, Beveled Dissecting Scissors
Each for $135.15
12-460-642
View Documents
Integra™ Miltex™
MH5126
17 cm Stainless Steel Curved, Beveled Dissecting Scissors
Each for $153.62
12-460-643
View Documents
Integra™ Miltex™
MH5128
22.9 cm Stainless Steel Straight, Beveled Dissecting Scissors
Each for $337.59
Description
Description
  • Stainless steel
  • Non-glare matted finish
  • Sharp/sharp scissors available in straight and curved patterns
  • Tungsten carbide inserts seamlessly bonded to upper and lower blades and feature gold ring handles
  • SuperCut scissors have a razor sharp edge that cuts effortlessly through tissue and lower blade has micro fine serrations to help prevent tissue slippage; black ring handles
Order Info
Cold or chemical sterilization not recommended for tungsten carbide scissors. SuperCut scissiors designed to cut tissue only; not recommended for use on suture material. Dropping or mishandling will cause the razor-sharp edge to nick or become irregular.
Specifications
Specifications
Dissecting
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'