Integra™ Miltex™ MeisterHand™ Operating Scissors
For general utility applications
Supplier: Integra™ Miltex™ MH52
Description
- Standard pattern
- Blunt-blunt and sharp blunt tips are ideal for cutting sutures, gauze, tubing and other materials
- Sharp-sharp tips are useful for cutting tendons and cartilage
- Stainless steel
- Non-glare matte finish
Specifications
Surgical Scissors | |
4.75 in. | |
Stainless Steel | |
Sharp/Sharp |
Straight | |
12.1 cm | |
Surgical, Precision |