Integra™ Miltex™ MeisterHand™ Operating Scissors
For general utility applications
$110.56 - $136.12
Specifications
Product Type | Surgical Scissors |
---|---|
Material | Stainless Steel |
For Use With (Application) | Surgical, Precision |
Catalog Number | Mfr. No. | Blade Style | Length (Metric) | Tip Style | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Blade Style | Length (Metric) | Tip Style | Price | Quantity | |||
12-460-636
|
Integra™ Miltex™
MH512 |
Straight | 12.1 cm | Sharp/Blunt |
Each for $110.56
|
|
|||
12-460-645
|
Integra™ Miltex™
MH516 |
Straight | 14.6 cm | Sharp/Blunt |
Each for $116.32
|
|
|||
12-460-644
|
Integra™ Miltex™
MH514 |
Straight | 12.7 cm | Sharp/Blunt |
Each for $114.82
|
|
|||
12-460-650
|
Integra™ Miltex™
MH52 |
Straight | 12.1 cm | Sharp/Sharp |
Each for $110.56
|
|
|||
12-460-651
|
Integra™ Miltex™
MH522 |
Straight | 12.1 cm | Blunt/Blunt |
Each for $118.03
|
|
|||
12-460-652
|
Integra™ Miltex™
MH526 |
Straight | 14.6 cm | Blunt/Blunt |
Each for $114.71
|
|
|||
12-460-668
|
Integra™ Miltex™
MH556 |
Curved | 13.97 cm | Blunt/Blunt |
Each for $130.50
|
|
|||
12-460-665
|
Integra™ Miltex™
MH536 |
Curved | 13.97 cm | Sharp/Sharp |
Each for $130.97
|
|
|||
12-460-666
|
Integra™ Miltex™
MH54 |
Straight | 12.7 cm | Sharp/Sharp |
Each for $113.15
|
|
|||
12-460-670
|
Integra™ Miltex™
MH56 |
Straight | 13.97 cm | Sharp/Sharp |
Each for $112.62
|
|
|||
12-460-671
|
Integra™ Miltex™
MH58 |
Straight | 16.51 cm | Sharp/Sharp |
Each for $136.12
|
|
|||
Description
- Standard pattern
- Blunt-blunt and sharp blunt tips are ideal for cutting sutures, gauze, tubing and other materials
- Sharp-sharp tips are useful for cutting tendons and cartilage
- Stainless steel
- Non-glare matte finish
Specifications
Surgical Scissors | |
Stainless Steel | |
Surgical, Precision |