Integra™ Miltex™ MeisterHand™ Operating Scissors
For general utility applications
Supplier:  Integra™ Miltex™ MH556
 View more versions of this product
Catalog No. 12-460-668
Explore available promotions
Description
Description
  • Standard pattern
  • Blunt-blunt and sharp blunt tips are ideal for cutting sutures, gauze, tubing and other materials
  • Sharp-sharp tips are useful for cutting tendons and cartilage
  • Stainless steel
  • Non-glare matte finish
Specifications
Specifications
Surgical Scissors
Standard
13.97 cm
Surgical, Precision
Curved
5.5 in.
Stainless Steel
Blunt/Blunt