missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Unisex Poly/Cotton Lab Coats
With front gripper closures
Supplier: Fisherbrand™ 19181574
Description
- Made of 65/35 Polycotton
- One outside left breast pocket, two outside lower pockets
- Standard pointed collar
- Front snap closure
- Straight cuffs
- White
Specifications
General Purpose Lab Coat | |
Men's | |
White | |
Open | |
Non-sterile | |
Standard Pointed |
3X-Large | |
Polycotton | |
Snaps | |
Reusable | |
3 | |
46.5 in. |