missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mercuric Nitrate, 0.100 N (N/10), Ricca Chemical

Supplier:  Ricca Chemical Company 473016

 View more versions of this product

Catalog No. 4730-16


Explore available promotions

Alert:

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.


Description

Description

Specifications

Specifications

Mercuric Nitrate
7783-34-8,7697-37-2,7732-18-5
MFCD00149736
KVICROHOONHSRH-UHFFFAOYSA-N
mercury(2+) hydrate dinitrate
3084029
500 mL
0.100Normal
Colorless
H2HgN2O7
mercuric nitrate monohydrate, unii-yj09edi5uy, mercury ii nitrate hydrate, yj09edi5uy, mercury 2+ ion hydrate dinitrate, nitric acid, mercury 2+ salt, monohydrate, mercury nitrate monohydrate, 324.60 anhydrous, mercury 2+ dinitrate hydrate, hgn2o6.xh2o x=1-2
O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
342.62
Amber Glass Bottle
Liquid

Chemical Identifiers

7783-34-8
342.62
KVICROHOONHSRH-UHFFFAOYSA-N
3084029
O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
H2HgN2O7
MFCD00149736
mercuric nitrate monohydrate, unii-yj09edi5uy, mercury ii nitrate hydrate, yj09edi5uy, mercury 2+ ion hydrate dinitrate, nitric acid, mercury 2+ salt, monohydrate, mercury nitrate monohydrate, 324.60 anhydrous, mercury 2+ dinitrate hydrate, hgn2o6.xh2o x=1-2
mercury(2+) hydrate dinitrate
Videos
SDS Documents

Documents

Promotions

Promotions