missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mercuric Nitrate, 0.100 N (N/10), Ricca Chemical
$178.63 - $280.16
Chemical Identifiers
CAS | 7783-34-8 |
---|---|
Molecular Formula | H2HgN2O7 |
Molecular Weight (g/mol) | 342.62 |
MDL Number | MFCD00149736 |
InChI Key | KVICROHOONHSRH-UHFFFAOYSA-N |
Synonym | mercuric nitrate monohydrate, unii-yj09edi5uy, mercury ii nitrate hydrate, yj09edi5uy, mercury 2+ ion hydrate dinitrate, nitric acid, mercury 2+ salt, monohydrate, mercury nitrate monohydrate, 324.60 anhydrous, mercury 2+ dinitrate hydrate, hgn2o6.xh2o x=1-2 |
PubChem CID | 3084029 |
IUPAC Name | mercury(2+) hydrate dinitrate |
SMILES | O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Chemical Identifiers
7783-34-8 | |
342.62 | |
KVICROHOONHSRH-UHFFFAOYSA-N | |
3084029 | |
O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
H2HgN2O7 | |
MFCD00149736 | |
mercuric nitrate monohydrate, unii-yj09edi5uy, mercury ii nitrate hydrate, yj09edi5uy, mercury 2+ ion hydrate dinitrate, nitric acid, mercury 2+ salt, monohydrate, mercury nitrate monohydrate, 324.60 anhydrous, mercury 2+ dinitrate hydrate, hgn2o6.xh2o x=1-2 | |
mercury(2+) hydrate dinitrate |
Specifications
Colorless | |
H2HgN2O7 | |
mercuric nitrate monohydrate, unii-yj09edi5uy, mercury ii nitrate hydrate, yj09edi5uy, mercury 2+ ion hydrate dinitrate, nitric acid, mercury 2+ salt, monohydrate, mercury nitrate monohydrate, 324.60 anhydrous, mercury 2+ dinitrate hydrate, hgn2o6.xh2o x=1-2 | |
O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
342.62 | |
Amber Glass Bottle | |
Liquid | |
Mercuric Nitrate |
7783-34-8,7697-37-2,7732-18-5 | |
MFCD00149736 | |
KVICROHOONHSRH-UHFFFAOYSA-N | |
mercury(2+) hydrate dinitrate | |
3084029 | |
500 mL | |
0.100Normal |