missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
meso-Hydrobenzoin, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2394614
Description
meso- Hydrobenzoin has been used in the preparation of trans-methyl meso- hydrobenzoin phosphite.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsmeso- Hydrobenzoin has been used in the preparation of trans-methyl meso- hydrobenzoin phosphite.
Solubility
Soluble in hot alcohol or chloroform.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Specifications
meso-Hydrobenzoin | |
134°C to 136°C | |
MFCD00064253 | |
2050814 | |
meso-hydrobenzoin, meso-1,2-diphenyl-1,2-ethanediol, unii-co9a49a84i, meso-stilbene glycol, 1r,2s-1,2-diphenylethane-1,2-diol, meso-1,2-diphenylethylene glycol, hydrobenzoin, meso, unii-q61g3433lb component | |
IHPDTPWNFBQHEB-UHFFFAOYNA-N | |
(1R,2S)-1,2-diphenylethane-1,2-diol | |
853018 | |
214.27 |
579-43-1 | |
C14H14O2 | |
25 g | |
14,4777 | |
Soluble in hot alcohol or chloroform. | |
OC(C(O)C1=CC=CC=C1)C1=CC=CC=C1 | |
214.26 | |
CHEBI:50015 | |
99% |
Chemical Identifiers
579-43-1 | |
214.26 | |
IHPDTPWNFBQHEB-UHFFFAOYNA-N | |
853018 | |
(1R,2S)-1,2-diphenylethane-1,2-diol |
C14H14O2 | |
MFCD00064253 | |
meso-hydrobenzoin, meso-1,2-diphenyl-1,2-ethanediol, unii-co9a49a84i, meso-stilbene glycol, 1r,2s-1,2-diphenylethane-1,2-diol, meso-1,2-diphenylethylene glycol, hydrobenzoin, meso, unii-q61g3433lb component | |
CHEBI:50015 | |
OC(C(O)C1=CC=CC=C1)C1=CC=CC=C1 |
Safety and Handling
EINECSNumber : 207-758-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only