missing translation for 'onlineSavingsMsg'
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits

meso-Hydrobenzoin, 95%, Thermo Scientific Chemicals

$126.11 - $483.52

Identifiants chimiques

CAS 579-43-1
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.26
MDL Number MFCD00064253
InChI Key IHPDTPWNFBQHEB-UHFFFAOYNA-N
Synonym meso-hydrobenzoin, meso-1,2-diphenyl-1,2-ethanediol, unii-co9a49a84i, meso-stilbene glycol, 1r,2s-1,2-diphenylethane-1,2-diol, meso-1,2-diphenylethylene glycol, hydrobenzoin, meso, unii-q61g3433lb component
PubChem CID 853018
ChEBI CHEBI:50015
IUPAC Name (1R,2S)-1,2-diphenylethane-1,2-diol
SMILES OC(C(O)C1=CC=CC=C1)C1=CC=CC=C1
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AAB2394606
Afficher les documents
Thermo Scientific Chemicals
B2394606
5 g
chaque for $126.11
 
AAB2394614
Afficher les documents
Thermo Scientific Chemicals
B2394614
25 g
chaque for $483.52
 
Description

Description

meso- Hydrobenzoin has been used in the preparation of trans-methyl meso- hydrobenzoin phosphite.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
meso- Hydrobenzoin has been used in the preparation of trans-methyl meso- hydrobenzoin phosphite.

Solubility
Soluble in hot alcohol or chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Spécifications

Identifiants chimiques

579-43-1
214.26
IHPDTPWNFBQHEB-UHFFFAOYNA-N
853018
(1R,2S)-1,2-diphenylethane-1,2-diol
C14H14O2
MFCD00064253
meso-hydrobenzoin, meso-1,2-diphenyl-1,2-ethanediol, unii-co9a49a84i, meso-stilbene glycol, 1r,2s-1,2-diphenylethane-1,2-diol, meso-1,2-diphenylethylene glycol, hydrobenzoin, meso, unii-q61g3433lb component
CHEBI:50015
OC(C(O)C1=CC=CC=C1)C1=CC=CC=C1

Spécifications

579-43-1
C14H14O2
5 g
14,4777
Soluble in hot alcohol or chloroform.
OC(C(O)C1=CC=CC=C1)C1=CC=CC=C1
214.26
CHEBI:50015
99%
134°C to 136°C
MFCD00064253
2050814
meso-hydrobenzoin, meso-1,2-diphenyl-1,2-ethanediol, unii-co9a49a84i, meso-stilbene glycol, 1r,2s-1,2-diphenylethane-1,2-diol, meso-1,2-diphenylethylene glycol, hydrobenzoin, meso, unii-q61g3433lb component
IHPDTPWNFBQHEB-UHFFFAOYNA-N
(1R,2S)-1,2-diphenylethane-1,2-diol
853018
214.27
meso-Hydrobenzoin
Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'einecsNumber' : 207-758-3

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only