missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MetFormin hydrochloride, 98%, For HPLC analysis, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0215169101

 View more versions of this product

Catalog No. ICN15169101

Explore available promotions

Description

Description

Specifications

Specifications

Metformin hydrochloride
1115-70-4
C4H12ClN5
1 g
OETHQSJEHLVLGH-UHFFFAOYSA-N
hydrogen 1-[(diaminomethylidene)amino]-N,N-dimethylmethanimidamide chloride
14219
0.98
For HPLC analysis
White
MFCD00012582
metformin hydrochloride, 1,1-dimethylbiguanide hydrochloride, metformin hcl, glucophage, riomet, diabefagos, meguan, metformin monohydrochloride, benofomin, denkaform
[H+].[Cl-].CN(C)C(=N)N=C(N)N
165.63
165.6
Crystalline Powder

Chemical Identifiers

1115-70-4
165.63
OETHQSJEHLVLGH-UHFFFAOYSA-N
14219
[H+].[Cl-].CN(C)C(=N)N=C(N)N
C4H12ClN5
MFCD00012582
metformin hydrochloride, 1,1-dimethylbiguanide hydrochloride, metformin hcl, glucophage, riomet, diabefagos, meguan, metformin monohydrochloride, benofomin, denkaform
hydrogen 1-[(diaminomethylidene)amino]-N,N-dimethylmethanimidamide chloride
SDS
Documents

Documents

Promotions

Promotions