missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methanol, ACS Reagent Grade, Absolute, Ricca Chemical
Supplier: Ricca Chemical Company 48291
Specifications
Methanol | |
67-56-1 | |
98.0 | |
4 L | |
Liquid | |
max. 0.0003 meq/g | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CO | |
32.04 | |
CHEBI:17790 | |
ACS |
Absolute | |
Colorless | |
100.0 | |
Natural Poly Bottle | |
max. 0.001% | |
CH4O | |
max. 0.1% | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
887 | |
≥99.8% |
Chemical Identifiers
67-56-1 | |
32.04 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
CHEBI:17790 | |
CO |
CH4O | |
MFCD00004595 | |
887 | |
methanol |
Safety and Handling
ShelfLife : 36 months