Learn More


Methanol (Certified ACS), Fisher Chemical™
Supplier: Fisher Chemical BPA41220
Specifications
Methanol | |
67-56-1 | |
0.791g/cm³ | |
20 L | |
Pass test | |
Liquid | |
Assay: ≥99.8% | |
0.0003meq/g max. | |
0.0002meq/g max. | |
128 hPa at 20°C | |
1230 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
887 | |
32.04 | |
Certified ACS Reagent |
-98°C | |
Colorless | |
64.7°C | |
12°C (53.6°F) | |
Steel Pail | |
10ppm max. | |
0.791 | |
CH4O | |
MFCD00004595 | |
0.55 cP at 20°C | |
0.1% Max. | |
Solubility in water: miscible | |
CO | |
32.04 | |
CHEBI:17790 | |
99.8% |
Chemical Identifiers
67-56-1 | |
32.04 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
887 | |
methanol |
CH4O | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CHEBI:17790 | |
CO |
Safety and Handling


Emergency Overview
Flammable liquid and vapor. Cannot be made non-poisonous. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Toxic: danger of very serious irreversible effects through inhalation, in contact with skin and if s. Explosive. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.
NFPA
Health:1
Flammability:3
Instability:0
Emergency Overview
Flammable liquid and vapor. Cannot be made non-poisonous. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Toxic: danger of ve
DANGER!
DOTInformation : DOT Class 3, : Flammable Liquid