Learn More
Methanol (Certified ACS), Fisher Chemical™
$203.63 - $6356.00
Chemical Identifiers
| CAS | 67-56-1 |
|---|---|
| Molecular Formula | CH4O |
| Molecular Weight (g/mol) | 32.04 |
| MDL Number | MFCD00004595 |
| InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
| Synonym | methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit |
| PubChem CID | 887 |
| ChEBI | CHEBI:17790 |
| IUPAC Name | methanol |
| SMILES | CO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
A412-500
|
Fisher Chemical
A412500 |
500 mL | HDPE Bottle |
Each for $204.18
Case of 6 Each for $1,225.08
|
|
||||
|
A412-1
|
Thermo Fisher Scientific
BPA4121 |
1 L | Glass Bottle |
Each for $343.73
Case of 6 Each for $2,062.38
|
|
||||
A412-4
|
Thermo Fisher Scientific
BPA4124 |
4 L | Glass Bottle |
Each for $203.63
Case of 4 Each for $814.52
|
|
||||
A412P-4
|
Fisher Chemical
BPA412P4 |
4 L | Poly Bottle |
Each for $252.73
Case of 4 Each for $1,010.92
|
|
||||
|
A412SK-4
|
Fisher Chemical
BPA412SK4 |
4 L | Safe-Cote™ Amber Glass |
Each for $445.73
Case of 4 Each for $1,782.92
|
|
||||
|
A412-20
|
Fisher Chemical
BPA41220 |
20 L | Steel Pail |
Each for $967.36
|
|
||||
|
A434-20
|
Thermo Fisher Scientific
BPA43420 |
20 L | Unichrome Steel Pail |
Each for $916.18
|
|
||||
|
A412-200
|
Thermo Fisher Scientific
BPA412200 |
200 L | Steel Drum |
Each for $6,356.00
|
|
||||
Chemical Identifiers
| 67-56-1 | |
| 32.04 | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| 887 | |
| methanol |
| CH4O | |
| MFCD00004595 | |
| methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
| CHEBI:17790 | |
| CO |
Specifications
| -98°C | |
| Colorless | |
| 64.7°C | |
| 12°C (53.6°F) | |
| HDPE Bottle | |
| 10ppm max. | |
| 0.791 | |
| CH4O | |
| MFCD00004595 | |
| 0.55 cP at 20°C | |
| 0.1% Max. | |
| Solubility in water: miscible | |
| CO | |
| 32.04 | |
| CHEBI:17790 | |
| 99.8% | |
| Methanol |
| 67-56-1 | |
| 0.791g/cm³ | |
| 500 mL | |
| Pass test | |
| Liquid | |
| Assay: ≥99.8% | |
| 0.0003meq/g max. | |
| 0.0002meq/g max. | |
| 128 hPa at 20°C | |
| 1230 | |
| methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| methanol | |
| 887 | |
| 32.04 | |
| Certified ACS Reagent |
Safety and Handling
Emergency Overview
Flammable liquid and vapor. Cannot be made non-poisonous. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Toxic: danger of very serious irreversible effects through inhalation, in contact with skin and if s. Explosive. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.
NFPA
Health:1
Flammability:3
Instability:0
Emergency Overview
Flammable liquid and vapor. Cannot be made non-poisonous. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Toxic: danger of ve
DANGER!
DOTInformation : DOT Class 3, : Flammable Liquid