missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 1-cyclopentene-1-carboxylate, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 397590100

 View more versions of this product

Catalog No. AC397590100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methyl 1-cyclopentene-1-carboxylate
White to Yellow
76°C to 78°C (9.0 mmHg)
Authentic
Glass bottle
C5H7COOCH3
10 g
methyl 1-cyclopentene-1-carboxylate, methyl cyclopent-1-enecarboxylate, methyl cyclopent-1-ene-1-carboxylate, cyclopentene-1-carboxylic acid methyl ester, methyl 1-cyclopentenecarboxylate, 1-cyclopentenecarboxylic acid methyl ester, 1-cyclopentene-1-carboxylic acid, methyl ester, methyl1-cyclopentene-1-carboxylate, 1-cyclopentene-1-carboxylic acid methyl ester, pubchem2487
COC(=O)C1=CCCC1
126.16
126.15
Crystalline Powder or Low Melting Solid
25662-28-6
1.0310g/mL
62°C
96% min. (GC)
C7H10O2
MFCD00239506
1.031
VTYCAXIAUKEGBQ-UHFFFAOYSA-N
methyl cyclopentene-1-carboxylate
549129
97%

Chemical Identifiers

25662-28-6
126.16
VTYCAXIAUKEGBQ-UHFFFAOYSA-N
549129
COC(=O)C1=CCCC1
C7H10O2
MFCD00239506
methyl 1-cyclopentene-1-carboxylate, methyl cyclopent-1-enecarboxylate, methyl cyclopent-1-ene-1-carboxylate, cyclopentene-1-carboxylic acid methyl ester, methyl 1-cyclopentenecarboxylate, 1-cyclopentenecarboxylic acid methyl ester, 1-cyclopentene-1-carboxylic acid, methyl ester, methyl1-cyclopentene-1-carboxylate, 1-cyclopentene-1-carboxylic acid methyl ester, pubchem2487
methyl cyclopentene-1-carboxylate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only