Learn More
Methyl 1-cyclopentene-1-carboxylate, 95%
CAS: 25662-28-6 | C7H10O2 | 126.16 g/mol
$381.01 - $381.01
Chemical Identifiers
| CAS | 25662-28-6 |
|---|---|
| Molecular Formula | C7H10O2 |
| Molecular Weight (g/mol) | 126.16 |
| MDL Number | MFCD00239506 |
| InChI Key | VTYCAXIAUKEGBQ-UHFFFAOYSA-N |
| Synonym | methyl 1-cyclopentene-1-carboxylate, methyl cyclopent-1-enecarboxylate, methyl cyclopent-1-ene-1-carboxylate, cyclopentene-1-carboxylic acid methyl ester, methyl 1-cyclopentenecarboxylate, 1-cyclopentenecarboxylic acid methyl ester, 1-cyclopentene-1-carboxylic acid, methyl ester, methyl1-cyclopentene-1-carboxylate, 1-cyclopentene-1-carboxylic acid methyl ester, pubchem2487 |
| PubChem CID | 549129 |
| IUPAC Name | methyl cyclopentene-1-carboxylate |
| SMILES | COC(=O)C1=CCCC1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC397590100
|
Thermo Scientific Chemicals
397590100 |
10 g | Glass bottle |
Each for $381.01
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 25662-28-6 | |
| 126.16 | |
| VTYCAXIAUKEGBQ-UHFFFAOYSA-N | |
| 549129 | |
| COC(=O)C1=CCCC1 |
| C7H10O2 | |
| MFCD00239506 | |
| methyl 1-cyclopentene-1-carboxylate, methyl cyclopent-1-enecarboxylate, methyl cyclopent-1-ene-1-carboxylate, cyclopentene-1-carboxylic acid methyl ester, methyl 1-cyclopentenecarboxylate, 1-cyclopentenecarboxylic acid methyl ester, 1-cyclopentene-1-carboxylic acid, methyl ester, methyl1-cyclopentene-1-carboxylate, 1-cyclopentene-1-carboxylic acid methyl ester, pubchem2487 | |
| methyl cyclopentene-1-carboxylate |
Specifications
| 25662-28-6 | |
| 1.0310g/mL | |
| 62°C | |
| 96% min. (GC) | |
| C7H10O2 | |
| MFCD00239506 | |
| 1.031 | |
| VTYCAXIAUKEGBQ-UHFFFAOYSA-N | |
| methyl cyclopentene-1-carboxylate | |
| 549129 | |
| 97% | |
| Methyl 1-cyclopentene-1-carboxylate |
| White to Yellow | |
| 76°C to 78°C (9.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| C5H7COOCH3 | |
| 10 g | |
| methyl 1-cyclopentene-1-carboxylate, methyl cyclopent-1-enecarboxylate, methyl cyclopent-1-ene-1-carboxylate, cyclopentene-1-carboxylic acid methyl ester, methyl 1-cyclopentenecarboxylate, 1-cyclopentenecarboxylic acid methyl ester, 1-cyclopentene-1-carboxylic acid, methyl ester, methyl1-cyclopentene-1-carboxylate, 1-cyclopentene-1-carboxylic acid methyl ester, pubchem2487 | |
| COC(=O)C1=CCCC1 | |
| 126.16 | |
| 126.15 | |
| Crystalline Powder or Low Melting Solid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
RUO – Research Use Only