missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 3,4-Dihydroxybenzoate 98.0+%, TCI America™
Supplier: TCI America M194325G
Specifications
Methyl 3,4-Dihydroxybenzoate | |
136°C | |
C8H8O4 | |
25 g | |
CUFLZUDASVUNOE-UHFFFAOYSA-N | |
methyl 3,4-dihydroxybenzoate | |
287064 | |
≥98.0% (GC,T) |
2150-43-8 | |
White | |
MFCD00016437 | |
methyl protocatechuate, protocatechuic acid, methyl ester, 3,4-dihydroxybenzoic acid methyl ester, protocatechuic acid methyl ester, benzoic acid, 3,4-dihydroxy-, methyl ester, 3,4-dihydroxymethylbenzoate, 3,4-dihydroxybenzoic acid methyl, mdhb, methyl 3,4-dihydroxy, acmc-1cfbz | |
COC(=O)C1=CC=C(O)C(O)=C1 | |
168.15 | |
168.15 | |
Crystalline Powder |
Chemical Identifiers
2150-43-8 | |
168.15 | |
CUFLZUDASVUNOE-UHFFFAOYSA-N | |
287064 | |
COC(=O)C1=CC=C(O)C(O)=C1 |
C8H8O4 | |
MFCD00016437 | |
methyl protocatechuate, protocatechuic acid, methyl ester, 3,4-dihydroxybenzoic acid methyl ester, protocatechuic acid methyl ester, benzoic acid, 3,4-dihydroxy-, methyl ester, 3,4-dihydroxymethylbenzoate, 3,4-dihydroxybenzoic acid methyl, mdhb, methyl 3,4-dihydroxy, acmc-1cfbz | |
methyl 3,4-dihydroxybenzoate |
Safety and Handling
TSCA : No