missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 3,4-Dihydroxybenzoate 98.0+%, TCI America™

$49.38 - $189.60

Chemical Identifiers

CAS 2150-43-8
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00016437
InChI Key CUFLZUDASVUNOE-UHFFFAOYSA-N
Synonym methyl protocatechuate, protocatechuic acid, methyl ester, 3,4-dihydroxybenzoic acid methyl ester, protocatechuic acid methyl ester, benzoic acid, 3,4-dihydroxy-, methyl ester, 3,4-dihydroxymethylbenzoate, 3,4-dihydroxybenzoic acid methyl, mdhb, methyl 3,4-dihydroxy, acmc-1cfbz
PubChem CID 287064
IUPAC Name methyl 3,4-dihydroxybenzoate
SMILES COC(=O)C1=CC=C(O)C(O)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M19435G
View Documents
TCI America
M19435G
5 g
Each for $49.38
 
M194325G
View Documents
TCI America
M194325G
25 g
Each for $189.60
 
Description

Description

Specifications

Chemical Identifiers

2150-43-8
168.15
CUFLZUDASVUNOE-UHFFFAOYSA-N
287064
COC(=O)C1=CC=C(O)C(O)=C1
C8H8O4
MFCD00016437
methyl protocatechuate, protocatechuic acid, methyl ester, 3,4-dihydroxybenzoic acid methyl ester, protocatechuic acid methyl ester, benzoic acid, 3,4-dihydroxy-, methyl ester, 3,4-dihydroxymethylbenzoate, 3,4-dihydroxybenzoic acid methyl, mdhb, methyl 3,4-dihydroxy, acmc-1cfbz
methyl 3,4-dihydroxybenzoate

Specifications

2150-43-8
White
MFCD00016437
methyl protocatechuate, protocatechuic acid, methyl ester, 3,4-dihydroxybenzoic acid methyl ester, protocatechuic acid methyl ester, benzoic acid, 3,4-dihydroxy-, methyl ester, 3,4-dihydroxymethylbenzoate, 3,4-dihydroxybenzoic acid methyl, mdhb, methyl 3,4-dihydroxy, acmc-1cfbz
COC(=O)C1=CC=C(O)C(O)=C1
168.15
168.15
Crystalline Powder
136°C
C8H8O4
5 g
CUFLZUDASVUNOE-UHFFFAOYSA-N
methyl 3,4-dihydroxybenzoate
287064
≥98.0% (GC,T)
Methyl 3,4-Dihydroxybenzoate
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'