missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 3-iodo-4-methylbenzoate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2873406

 View more versions of this product

Catalog No. AAH2873406

Explore available promotions

Description

Description

Methyl 3-Iodo-4-methylbenzoate is used as a reagent in the synthesis of [(pyrazolo[1,5-a]pyridmidinyl)ethynyl]benzamides as selective discoidin domain receptor 1 (DDR1) inhibitors. Also used as a reagent in the preparation of orally bioavailable GZD824 targeting Breakpoint Cluster Region-Abelson (Bcr-Abl) kinase and overcoming antitumor drug resistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-Iodo-4-methylbenzoate is used as a reagent in the synthesis of [(pyrazolo[1,5-a]pyridmidinyl)ethynyl]benzamides as selective discoidin domain receptor 1 (DDR1) inhibitors. Also used as a reagent in the preparation of orally bioavailable GZD824 targeting Breakpoint Cluster Region-Abelson (Bcr-Abl) kinase and overcoming antitumor drug resistance.

Solubility
Not miscible or difficult to mix with water.

Notes
Light Sensitive, store in dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

Methyl 3-iodo-4-methylbenzoate
C9H9IO2
MFCD00230583
Light sensitive
Not miscible or difficult to mix with water.
CC1=C(C=C(C=C1)C(=O)OC)I
276.073
276.07
90347-66-3
1.5975
5 g
3-iodo-4-methylbenzoic acid methyl ester, methyl 3-iodo-p-toluate, methyl-3-iodo-4-methylbenzoate, methyl 3-iodo-4-methyl-benzoate, 2-iodo-4-methoxycarbonyl toluene, benzoic acid, 3-iodo-4-methyl-, methyl ester, methyl3-iodo-4-methylbenzoate, 3-iodo-4-methylbenzoic acidmethyl ester, methyl 3-iodo-4-meth, ksc495m1r
NKMHAOTZPFVSPC-UHFFFAOYSA-N
methyl 3-iodo-4-methylbenzoate
13614033
98%

Chemical Identifiers

90347-66-3
276.073
NKMHAOTZPFVSPC-UHFFFAOYSA-N
13614033
CC1=C(C=C(C=C1)C(=O)OC)I
C9H9IO2
MFCD00230583
3-iodo-4-methylbenzoic acid methyl ester, methyl 3-iodo-p-toluate, methyl-3-iodo-4-methylbenzoate, methyl 3-iodo-4-methyl-benzoate, 2-iodo-4-methoxycarbonyl toluene, benzoic acid, 3-iodo-4-methyl-, methyl ester, methyl3-iodo-4-methylbenzoate, 3-iodo-4-methylbenzoic acidmethyl ester, methyl 3-iodo-4-meth, ksc495m1r
methyl 3-iodo-4-methylbenzoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only