Learn More
Methyl 3-iodo-4-methylbenzoate, 98%, Thermo Scientific Chemicals
$112.32 - $315.93
Chemical Identifiers
CAS | 90347-66-3 |
---|---|
Molecular Formula | C9H9IO2 |
Molecular Weight (g/mol) | 276.073 |
MDL Number | MFCD00230583 |
InChI Key | NKMHAOTZPFVSPC-UHFFFAOYSA-N |
Synonym | 3-iodo-4-methylbenzoic acid methyl ester, methyl 3-iodo-p-toluate, methyl-3-iodo-4-methylbenzoate, methyl 3-iodo-4-methyl-benzoate, 2-iodo-4-methoxycarbonyl toluene, benzoic acid, 3-iodo-4-methyl-, methyl ester, methyl3-iodo-4-methylbenzoate, 3-iodo-4-methylbenzoic acidmethyl ester, methyl 3-iodo-4-meth, ksc495m1r |
PubChem CID | 13614033 |
IUPAC Name | methyl 3-iodo-4-methylbenzoate |
SMILES | CC1=C(C=C(C=C1)C(=O)OC)I |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH2873406
|
Thermo Scientific Chemicals
H2873406 |
5 g |
N/A
|
|
||||||
AAH2873414
|
Thermo Scientific Chemicals
H2873414 |
25 g |
N/A
|
|
||||||
Description
Methyl 3-Iodo-4-methylbenzoate is used as a reagent in the synthesis of [(pyrazolo[1,5-a]pyridmidinyl)ethynyl]benzamides as selective discoidin domain receptor 1 (DDR1) inhibitors. Also used as a reagent in the preparation of orally bioavailable GZD824 targeting Breakpoint Cluster Region-Abelson (Bcr-Abl) kinase and overcoming antitumor drug resistance.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 3-Iodo-4-methylbenzoate is used as a reagent in the synthesis of [(pyrazolo[1,5-a]pyridmidinyl)ethynyl]benzamides as selective discoidin domain receptor 1 (DDR1) inhibitors. Also used as a reagent in the preparation of orally bioavailable GZD824 targeting Breakpoint Cluster Region-Abelson (Bcr-Abl) kinase and overcoming antitumor drug resistance.
Solubility
Not miscible or difficult to mix with water.
Notes
Light Sensitive, store in dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
90347-66-3 | |
276.073 | |
NKMHAOTZPFVSPC-UHFFFAOYSA-N | |
13614033 | |
CC1=C(C=C(C=C1)C(=O)OC)I |
C9H9IO2 | |
MFCD00230583 | |
3-iodo-4-methylbenzoic acid methyl ester, methyl 3-iodo-p-toluate, methyl-3-iodo-4-methylbenzoate, methyl 3-iodo-4-methyl-benzoate, 2-iodo-4-methoxycarbonyl toluene, benzoic acid, 3-iodo-4-methyl-, methyl ester, methyl3-iodo-4-methylbenzoate, 3-iodo-4-methylbenzoic acidmethyl ester, methyl 3-iodo-4-meth, ksc495m1r | |
methyl 3-iodo-4-methylbenzoate |
Specifications
90347-66-3 | |
1.5975 | |
5 g | |
3-iodo-4-methylbenzoic acid methyl ester, methyl 3-iodo-p-toluate, methyl-3-iodo-4-methylbenzoate, methyl 3-iodo-4-methyl-benzoate, 2-iodo-4-methoxycarbonyl toluene, benzoic acid, 3-iodo-4-methyl-, methyl ester, methyl3-iodo-4-methylbenzoate, 3-iodo-4-methylbenzoic acidmethyl ester, methyl 3-iodo-4-meth, ksc495m1r | |
NKMHAOTZPFVSPC-UHFFFAOYSA-N | |
methyl 3-iodo-4-methylbenzoate | |
13614033 | |
98% |
C9H9IO2 | |
MFCD00230583 | |
Light sensitive | |
Not miscible or difficult to mix with water. | |
CC1=C(C=C(C=C1)C(=O)OC)I | |
276.073 | |
276.07 | |
Methyl 3-iodo-4-methylbenzoate |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only