missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-(chloromethyl)benzoate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 391310010

 View more versions of this product

Catalog No. AC391310010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Methyl 4-(chloromethyl)benzoate
160.0°C to 165.0°C (30.0mmHg)
96% min. (GC)
C9H9ClO2
MFCD00511224
methyl 4-chloromethyl benzoate, 4-chloromethyl benzoic acid methyl ester, methyl-4-chloromethylbenzoate, benzoic acid, 4-chloromethyl-, methyl ester, 4-methoxycarbonyl benzyl chloride, methyl alpha-chloro-p-toluate, methyl p-chloromethyl benzoate, p-methoxycarbonyl benzyl chloride, methyl a-chloro-p-toluate, p-toluic acid, .alpha.-chloro-, methyl ester
COC(=O)C1=CC=C(C=C1)CCl
184.62
184.62
34040-64-7
Authentic
Glass bottle
ClCH2C6H4C(O)OCH3
1g
SATDLKYRVXFXRE-UHFFFAOYSA-N
methyl 4-(chloromethyl)benzoate
36664
97%

Chemical Identifiers

34040-64-7
184.62
SATDLKYRVXFXRE-UHFFFAOYSA-N
36664
COC(=O)C1=CC=C(C=C1)CCl
C9H9ClO2
MFCD00511224
methyl 4-chloromethyl benzoate, 4-chloromethyl benzoic acid methyl ester, methyl-4-chloromethylbenzoate, benzoic acid, 4-chloromethyl-, methyl ester, 4-methoxycarbonyl benzyl chloride, methyl alpha-chloro-p-toluate, methyl p-chloromethyl benzoate, p-methoxycarbonyl benzyl chloride, methyl a-chloro-p-toluate, p-toluic acid, .alpha.-chloro-, methyl ester
methyl 4-(chloromethyl)benzoate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 251-806-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only