Learn More
Methyl 4-(chloromethyl)benzoate, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 34040-64-7 |
---|---|
Molecular Formula | C9H9ClO2 |
Molecular Weight (g/mol) | 184.62 |
MDL Number | MFCD00511224 |
InChI Key | SATDLKYRVXFXRE-UHFFFAOYSA-N |
Synonym | methyl 4-chloromethyl benzoate, 4-chloromethyl benzoic acid methyl ester, methyl-4-chloromethylbenzoate, benzoic acid, 4-chloromethyl-, methyl ester, 4-methoxycarbonyl benzyl chloride, methyl alpha-chloro-p-toluate, methyl p-chloromethyl benzoate, p-methoxycarbonyl benzyl chloride, methyl a-chloro-p-toluate, p-toluic acid, .alpha.-chloro-, methyl ester |
PubChem CID | 36664 |
IUPAC Name | methyl 4-(chloromethyl)benzoate |
SMILES | COC(=O)C1=CC=C(C=C1)CCl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC391310010
|
Thermo Scientific Chemicals
391310010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
34040-64-7 | |
184.62 | |
SATDLKYRVXFXRE-UHFFFAOYSA-N | |
36664 | |
COC(=O)C1=CC=C(C=C1)CCl |
C9H9ClO2 | |
MFCD00511224 | |
methyl 4-chloromethyl benzoate, 4-chloromethyl benzoic acid methyl ester, methyl-4-chloromethylbenzoate, benzoic acid, 4-chloromethyl-, methyl ester, 4-methoxycarbonyl benzyl chloride, methyl alpha-chloro-p-toluate, methyl p-chloromethyl benzoate, p-methoxycarbonyl benzyl chloride, methyl a-chloro-p-toluate, p-toluic acid, .alpha.-chloro-, methyl ester | |
methyl 4-(chloromethyl)benzoate |
Specifications
34040-64-7 | |
Authentic | |
Glass bottle | |
ClCH2C6H4C(O)OCH3 | |
1g | |
SATDLKYRVXFXRE-UHFFFAOYSA-N | |
methyl 4-(chloromethyl)benzoate | |
36664 | |
97% |
160.0°C to 165.0°C (30.0mmHg) | |
96% min. (GC) | |
C9H9ClO2 | |
MFCD00511224 | |
methyl 4-chloromethyl benzoate, 4-chloromethyl benzoic acid methyl ester, methyl-4-chloromethylbenzoate, benzoic acid, 4-chloromethyl-, methyl ester, 4-methoxycarbonyl benzyl chloride, methyl alpha-chloro-p-toluate, methyl p-chloromethyl benzoate, p-methoxycarbonyl benzyl chloride, methyl a-chloro-p-toluate, p-toluic acid, .alpha.-chloro-, methyl ester | |
COC(=O)C1=CC=C(C=C1)CCl | |
184.62 | |
184.62 | |
Methyl 4-(chloromethyl)benzoate |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 251-806-6
RUO â Research Use Only