missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-hydroxy-3,5-dimethoxybenzoate, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1805406

 View more versions of this product

Catalog No. AAA1805406



Description

Description

The scheme of laccase-catalysed iodide oxidation includes stadium of MS interaction with oxidized laccase with concomitant production of MS(ox). Thereaction of MS(ox) with iodide produced triiodide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The scheme of laccase-catalysed iodide oxidation includes stadium of MS interaction with oxidized laccase with concomitant production of MS(ox). Thereaction of MS(ox) with iodide produced triiodide.

Solubility
Soluble in alcohol, ether, slightly soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

Methyl 4-hydroxy-3,5-dimethoxybenzoate
102°C to 108°C
MFCD00017199
2217524
Soluble in alcohol,ether,slightly soluble in water.
COC1=CC(=CC(=C1O)OC)C(=O)OC
212.201
CHEBI:45820
≥98%
884-35-5
C10H12O5
5 g
methyl syringate, methyl 3,5-dimethoxy-4-hydroxybenzoate, benzoic acid, 4-hydroxy-3,5-dimethoxy-, methyl ester, syringic acid methyl ester, methyl 4-hydroxy-3,5-dimethoxy-benzoate, acmc-1bjig, ksc496o5n, syringic acid monomethyl ester, rarechem al bf 0102, timtec-bb sbb016976
ZMXJAEGJWHJMGX-UHFFFAOYSA-N
methyl 4-hydroxy-3,5-dimethoxybenzoate
70164
212.2

Chemical Identifiers

884-35-5
212.201
ZMXJAEGJWHJMGX-UHFFFAOYSA-N
70164
methyl 4-hydroxy-3,5-dimethoxybenzoate
C10H12O5
MFCD00017199
methyl syringate, methyl 3,5-dimethoxy-4-hydroxybenzoate, benzoic acid, 4-hydroxy-3,5-dimethoxy-, methyl ester, syringic acid methyl ester, methyl 4-hydroxy-3,5-dimethoxy-benzoate, acmc-1bjig, ksc496o5n, syringic acid monomethyl ester, rarechem al bf 0102, timtec-bb sbb016976
CHEBI:45820
COC1=CC(=CC(=C1O)OC)C(=O)OC
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 429-050-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only