Learn More
Methyl 4-hydroxy-3,5-dimethoxybenzoate, 98+%, Thermo Scientific Chemicals
$98.82 - $286.89
Chemical Identifiers
CAS | 884-35-5 |
---|---|
Molecular Formula | C10H12O5 |
Molecular Weight (g/mol) | 212.201 |
MDL Number | MFCD00017199 |
InChI Key | ZMXJAEGJWHJMGX-UHFFFAOYSA-N |
Synonym | methyl syringate, methyl 3,5-dimethoxy-4-hydroxybenzoate, benzoic acid, 4-hydroxy-3,5-dimethoxy-, methyl ester, syringic acid methyl ester, methyl 4-hydroxy-3,5-dimethoxy-benzoate, acmc-1bjig, ksc496o5n, syringic acid monomethyl ester, rarechem al bf 0102, timtec-bb sbb016976 |
PubChem CID | 70164 |
ChEBI | CHEBI:45820 |
IUPAC Name | methyl 4-hydroxy-3,5-dimethoxybenzoate |
SMILES | COC1=CC(=CC(=C1O)OC)C(=O)OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1805406
|
Thermo Scientific Chemicals
A1805406 |
5 g |
Each for $98.82
|
|
|||||
AAA1805414
|
Thermo Scientific Chemicals
A1805414 |
25 g |
Each for $286.89
|
|
|||||
Description
The scheme of laccase-catalysed iodide oxidation includes stadium of MS interaction with oxidized laccase with concomitant production of MS(ox). Thereaction of MS(ox) with iodide produced triiodide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThe scheme of laccase-catalysed iodide oxidation includes stadium of MS interaction with oxidized laccase with concomitant production of MS(ox). Thereaction of MS(ox) with iodide produced triiodide.
Solubility
Soluble in alcohol, ether, slightly soluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
884-35-5 | |
212.201 | |
ZMXJAEGJWHJMGX-UHFFFAOYSA-N | |
70164 | |
methyl 4-hydroxy-3,5-dimethoxybenzoate |
C10H12O5 | |
MFCD00017199 | |
methyl syringate, methyl 3,5-dimethoxy-4-hydroxybenzoate, benzoic acid, 4-hydroxy-3,5-dimethoxy-, methyl ester, syringic acid methyl ester, methyl 4-hydroxy-3,5-dimethoxy-benzoate, acmc-1bjig, ksc496o5n, syringic acid monomethyl ester, rarechem al bf 0102, timtec-bb sbb016976 | |
CHEBI:45820 | |
COC1=CC(=CC(=C1O)OC)C(=O)OC |
Specifications
884-35-5 | |
C10H12O5 | |
5 g | |
methyl syringate, methyl 3,5-dimethoxy-4-hydroxybenzoate, benzoic acid, 4-hydroxy-3,5-dimethoxy-, methyl ester, syringic acid methyl ester, methyl 4-hydroxy-3,5-dimethoxy-benzoate, acmc-1bjig, ksc496o5n, syringic acid monomethyl ester, rarechem al bf 0102, timtec-bb sbb016976 | |
ZMXJAEGJWHJMGX-UHFFFAOYSA-N | |
methyl 4-hydroxy-3,5-dimethoxybenzoate | |
70164 | |
212.2 | |
Methyl 4-hydroxy-3,5-dimethoxybenzoate |
102°C to 108°C | |
MFCD00017199 | |
2217524 | |
Soluble in alcohol,ether,slightly soluble in water. | |
COC1=CC(=CC(=C1O)OC)C(=O)OC | |
212.201 | |
CHEBI:45820 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 429-050-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only