missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-hydroxy-6-methyl-2-oxo-3-cyclohexene-1-carboxylate, 99%, Thermo Scientific Chemicals

$128.58 - $128.58

Chemical Identifiers

CAS 39493-62-4
Molecular Formula C9H12O4
Molecular Weight (g/mol) 184.191
MDL Number MFCD01075661
InChI Key SEBIQMPNDQLYOX-UHFFFAOYSA-N
Synonym methyl 4-hydroxy-6-methyl-2-oxo-3-cyclohexene-1-carboxylate, methyl 4-hydroxy-6-methyl-2-oxocyclohex-3-ene-1-carboxylate, methyl 2-hydroxy-6-methyl-4-oxocyclohex-2-enecarboxylate, methyl 4-hydroxy-6-methyl-2-oxocyclohex-3-enecarboxylate, acmc-20akgd, methyl 4-hydroxy-6-methyl-2-oxo-cyclohex-3-en-1-oate, methyl 2-hydroxy-6-methyl-4-oxo-2-cyclohexene-1-carboxylate, 3-cyclohexene-1-carboxylicacid, 4-hydroxy-6-methyl-2-oxo-, methyl ester, 6-methyl-2-hydroxy-4-oxo-2-cyclohexene-1-carboxylic acid methyl ester
PubChem CID 3766543
IUPAC Name methyl 2-hydroxy-6-methyl-4-oxocyclohex-2-ene-1-carboxylate
SMILES CC1CC(=O)C=C(C1C(=O)OC)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1462206
View Documents
Thermo Scientific Chemicals
L1462206
5 g
Each for $128.58
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

39493-62-4
184.191
SEBIQMPNDQLYOX-UHFFFAOYSA-N
3766543
CC1CC(=O)C=C(C1C(=O)OC)O
C9H12O4
MFCD01075661
methyl 4-hydroxy-6-methyl-2-oxo-3-cyclohexene-1-carboxylate, methyl 4-hydroxy-6-methyl-2-oxocyclohex-3-ene-1-carboxylate, methyl 2-hydroxy-6-methyl-4-oxocyclohex-2-enecarboxylate, methyl 4-hydroxy-6-methyl-2-oxocyclohex-3-enecarboxylate, acmc-20akgd, methyl 4-hydroxy-6-methyl-2-oxo-cyclohex-3-en-1-oate, methyl 2-hydroxy-6-methyl-4-oxo-2-cyclohexene-1-carboxylate, 3-cyclohexene-1-carboxylicacid, 4-hydroxy-6-methyl-2-oxo-, methyl ester, 6-methyl-2-hydroxy-4-oxo-2-cyclohexene-1-carboxylic acid methyl ester
methyl 2-hydroxy-6-methyl-4-oxocyclohex-2-ene-1-carboxylate

Specifications

39493-62-4
C9H12O4
5 g
methyl 4-hydroxy-6-methyl-2-oxo-3-cyclohexene-1-carboxylate, methyl 4-hydroxy-6-methyl-2-oxocyclohex-3-ene-1-carboxylate, methyl 2-hydroxy-6-methyl-4-oxocyclohex-2-enecarboxylate, methyl 4-hydroxy-6-methyl-2-oxocyclohex-3-enecarboxylate, acmc-20akgd, methyl 4-hydroxy-6-methyl-2-oxo-cyclohex-3-en-1-oate, methyl 2-hydroxy-6-methyl-4-oxo-2-cyclohexene-1-carboxylate, 3-cyclohexene-1-carboxylicacid, 4-hydroxy-6-methyl-2-oxo-, methyl ester, 6-methyl-2-hydroxy-4-oxo-2-cyclohexene-1-carboxylic acid methyl ester
CC1CC(=O)C=C(C1C(=O)OC)O
184.191
184.19
Methyl 4-hydroxy-6-methyl-2-oxo-3-cyclohexene-1-carboxylate
122°C to 124°C
MFCD01075661
5258501
SEBIQMPNDQLYOX-UHFFFAOYSA-N
methyl 2-hydroxy-6-methyl-4-oxocyclohex-2-ene-1-carboxylate
3766543
99%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only