missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl cyanoacetate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 148642500

 View more versions of this product

Catalog No. AC148642500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Methyl cyanoacetate
105-34-0
204°C to 207°C
Authentic
Glass bottle
1.4170 to 1.419
MFCD00001939
02, 584
methyl cyanoacetate, cyanoacetic acid methyl ester, methyl cyanoethanoate, acetic acid, cyano-, methyl ester, usaf kf-22, methylester kyseliny kyanoctove, methylcyanoacetate, malonic methyl ester nitrile, acetic acid, 2-cyano-, methyl ester, methyl cyano acetate
ANGDWNBGPBMQHW-UHFFFAOYSA-N
methyl 2-cyanoacetate
7747
99.09
97%
1.1200g/mL
>110°C
96% min. (GC)
C4H5NO2
NCCH2CO2CH3
250mL
1.12
Solubility in water: 54g/L in water (20°C). Other solubilities: soluble in alcohol and ether
COC(=O)CC#N
99.09
CHEBI:51928
97%

Chemical Identifiers

105-34-0
99.09
ANGDWNBGPBMQHW-UHFFFAOYSA-N
7747
methyl 2-cyanoacetate
C4H5NO2
MFCD00001939
methyl cyanoacetate, cyanoacetic acid methyl ester, methyl cyanoethanoate, acetic acid, cyano-, methyl ester, usaf kf-22, methylester kyseliny kyanoctove, methylcyanoacetate, malonic methyl ester nitrile, acetic acid, 2-cyano-, methyl ester, methyl cyano acetate
CHEBI:51928
COC(=O)CC#N
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-288-8

RTECSNumber : AG4375000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only