missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl cyanoacetate, 97%, Thermo Scientific™

Chemical Identifiers

CAS 105-34-0
Molecular Formula C4H5NO2
Molecular Weight (g/mol) 99.09
MDL Number MFCD00001939
InChI Key ANGDWNBGPBMQHW-UHFFFAOYSA-N
Synonym methyl cyanoacetate, cyanoacetic acid methyl ester, methyl cyanoethanoate, acetic acid, cyano-, methyl ester, usaf kf-22, methylester kyseliny kyanoctove, methylcyanoacetate, malonic methyl ester nitrile, acetic acid, 2-cyano-, methyl ester, methyl cyano acetate
PubChem CID 7747
ChEBI CHEBI:51928
IUPAC Name methyl 2-cyanoacetate
SMILES COC(=O)CC#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC148642500
SDS View Documents
Thermo Scientific Chemicals
148642500
250mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC148640010
SDS View Documents
Thermo Scientific Chemicals
148640010
1L Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

105-34-0
99.09
ANGDWNBGPBMQHW-UHFFFAOYSA-N
7747
methyl 2-cyanoacetate
C4H5NO2
MFCD00001939
methyl cyanoacetate, cyanoacetic acid methyl ester, methyl cyanoethanoate, acetic acid, cyano-, methyl ester, usaf kf-22, methylester kyseliny kyanoctove, methylcyanoacetate, malonic methyl ester nitrile, acetic acid, 2-cyano-, methyl ester, methyl cyano acetate
CHEBI:51928
COC(=O)CC#N

Specifications

105-34-0
204°C to 207°C
Authentic
Glass bottle
1.4170 to 1.419
MFCD00001939
02, 584
methyl cyanoacetate, cyanoacetic acid methyl ester, methyl cyanoethanoate, acetic acid, cyano-, methyl ester, usaf kf-22, methylester kyseliny kyanoctove, methylcyanoacetate, malonic methyl ester nitrile, acetic acid, 2-cyano-, methyl ester, methyl cyano acetate
ANGDWNBGPBMQHW-UHFFFAOYSA-N
methyl 2-cyanoacetate
7747
99.09
Methyl cyanoacetate, 97%
1.1200g/mL
>110°C
96% min. (GC)
C4H5NO2
NCCH2CO2CH3
250mL
1.12
Solubility in water: 54g/L in water (20°C). Other solubilities: soluble in alcohol and ether
COC(=O)CC#N
99.09
CHEBI:51928
97%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-288-8

RTECSNumber : AG4375000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only