missing translation for 'onlineSavingsMsg'
Learn More

Methyl cyclopentanecarboxylate, 97%

CAS: 4630-80-2 | C7H12O2 | 128.17 g/mol

Supplier:  Thermo Scientific Chemicals L0074106

 View more versions of this product

Catalog No. AAL0074106

Only null left
Explore available promotions

Description

Description

Methyl cyclopentanecarboxylate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl cyclopentanecarboxylate

Solubility
Difficult to mix.

Notes
Protect from heat. Store away from oxidizing agents.
Specifications

Specifications

Methyl cyclopentanecarboxylate
81°C to 82°C (5 mmHg)
1.436
5 g
cyclopentanecarboxylic acid, methyl ester, cyclopentanecarboxylic acid methyl ester, methyl cyclopentanoate, acmc-1am5n, methylcyclopentanecarboxylate, methylcyclopentane carboxylate, methyl cyclopentane carboxylate, methyl-1-cyclopentanecarboxylate, cyclopentanoic acid methyl ester, cyclopentanecarboxylic acid methyl
IIHIJFJSXPDTNO-UHFFFAOYSA-N
methyl cyclopentanecarboxylate
78365
98%
4630-80-2
C7H12O2
MFCD00019290
2040762
Difficult to mix.
COC(=O)C1CCCC1
128.17
128.17

Chemical Identifiers

4630-80-2
128.17
IIHIJFJSXPDTNO-UHFFFAOYSA-N
78365
COC(=O)C1CCCC1
C7H12O2
MFCD00019290
cyclopentanecarboxylic acid, methyl ester, cyclopentanecarboxylic acid methyl ester, methyl cyclopentanoate, acmc-1am5n, methylcyclopentanecarboxylate, methylcyclopentane carboxylate, methyl cyclopentane carboxylate, methyl-1-cyclopentanecarboxylate, cyclopentanoic acid methyl ester, cyclopentanecarboxylic acid methyl
methyl cyclopentanecarboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P330-P374-P380-P501c

H226-H302

DOTInformation : Hazard Class: 3; Packaging Group: III

EINECSNumber : 225-049-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only