missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl cyclopentanecarboxylate, 97%, Thermo Scientific Chemicals
$160.88 - $582.55
Chemical Identifiers
CAS | 4630-80-2 |
---|---|
Molecular Formula | C7H12O2 |
Molecular Weight (g/mol) | 128.17 |
MDL Number | MFCD00019290 |
InChI Key | IIHIJFJSXPDTNO-UHFFFAOYSA-N |
Synonym | cyclopentanecarboxylic acid, methyl ester, cyclopentanecarboxylic acid methyl ester, methyl cyclopentanoate, acmc-1am5n, methylcyclopentanecarboxylate, methylcyclopentane carboxylate, methyl cyclopentane carboxylate, methyl-1-cyclopentanecarboxylate, cyclopentanoic acid methyl ester, cyclopentanecarboxylic acid methyl |
PubChem CID | 78365 |
IUPAC Name | methyl cyclopentanecarboxylate |
SMILES | COC(=O)C1CCCC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL0074106
|
Thermo Scientific Chemicals
L0074106 |
5 g |
Each for $160.88
|
|
||||||
AAL0074114
|
Thermo Scientific Chemicals
L0074114 |
25 g |
Each for $582.55
|
|
||||||
Description
Methyl cyclopentanecarboxylate
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl cyclopentanecarboxylate
Solubility
Difficult to mix.
Notes
Protect from heat. Store away from oxidizing agents.
Chemical Identifiers
4630-80-2 | |
128.17 | |
IIHIJFJSXPDTNO-UHFFFAOYSA-N | |
78365 | |
COC(=O)C1CCCC1 |
C7H12O2 | |
MFCD00019290 | |
cyclopentanecarboxylic acid, methyl ester, cyclopentanecarboxylic acid methyl ester, methyl cyclopentanoate, acmc-1am5n, methylcyclopentanecarboxylate, methylcyclopentane carboxylate, methyl cyclopentane carboxylate, methyl-1-cyclopentanecarboxylate, cyclopentanoic acid methyl ester, cyclopentanecarboxylic acid methyl | |
methyl cyclopentanecarboxylate |
Specifications
4630-80-2 | |
C7H12O2 | |
MFCD00019290 | |
2040762 | |
Difficult to mix. | |
COC(=O)C1CCCC1 | |
128.17 | |
128.17 | |
Methyl cyclopentanecarboxylate |
81°C to 82°C (5 mmHg) | |
1.436 | |
5 g | |
cyclopentanecarboxylic acid, methyl ester, cyclopentanecarboxylic acid methyl ester, methyl cyclopentanoate, acmc-1am5n, methylcyclopentanecarboxylate, methylcyclopentane carboxylate, methyl cyclopentane carboxylate, methyl-1-cyclopentanecarboxylate, cyclopentanoic acid methyl ester, cyclopentanecarboxylic acid methyl | |
IIHIJFJSXPDTNO-UHFFFAOYSA-N | |
methyl cyclopentanecarboxylate | |
78365 | |
98% |
Safety and Handling
P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P330-P374-P380-P501c
H226-H302
DOTInformation : Hazard Class: 3; Packaging Group: III
EINECSNumber : 225-049-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only