missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl eicosanoate, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04239904

 View more versions of this product

Catalog No. AA4239904


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methyl eicosanoate
48°C
215°C (10 mmHg)
C21H42O2
2 g
Soluble in hot alcohol. Sparingly Soluble in water.
CCCCCCCCCCCCCCCCCCCC(=O)OC
326.57
326.56
Crystalline Powder
1120-28-1
White
>110°C (230°F)
MFCD00009014
methyl arachidate, eicosanoic acid, methyl ester, arachidic acid methyl ester, methyl eicosanoate, methyl arachisate, arachidic acid methylester, eicosanoic acid methyl ester, methylarachidate, methyl aracidate
QGBRLVONZXHAKJ-UHFFFAOYSA-N
methyl icosanoate
14259
≥98%

Chemical Identifiers

1120-28-1
326.57
QGBRLVONZXHAKJ-UHFFFAOYSA-N
14259
CCCCCCCCCCCCCCCCCCCC(=O)OC
C21H42O2
MFCD00009014
methyl arachidate, eicosanoic acid, methyl ester, arachidic acid methyl ester, methyl eicosanoate, methyl arachisate, arachidic acid methylester, eicosanoic acid methyl ester, methylarachidate, methyl aracidate
methyl icosanoate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 214-304-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only