missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl eicosanoate, 98+%, Thermo Scientific Chemicals

$129.47 - $481.60

Chemical Identifiers

CAS 1120-28-1
Molecular Formula C21H42O2
Molecular Weight (g/mol) 326.57
MDL Number MFCD00009014
InChI Key QGBRLVONZXHAKJ-UHFFFAOYSA-N
Synonym methyl arachidate, eicosanoic acid, methyl ester, arachidic acid methyl ester, methyl eicosanoate, methyl arachisate, arachidic acid methylester, eicosanoic acid methyl ester, methylarachidate, methyl aracidate
PubChem CID 14259
IUPAC Name methyl icosanoate
SMILES CCCCCCCCCCCCCCCCCCCC(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4239904
View Documents
Thermo Scientific Chemicals
04239904
2 g
Each for $129.47
 
AA4239909
View Documents
Thermo Scientific Chemicals
04239909
10 g
Each for $481.60
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1120-28-1
326.57
QGBRLVONZXHAKJ-UHFFFAOYSA-N
14259
CCCCCCCCCCCCCCCCCCCC(=O)OC
C21H42O2
MFCD00009014
methyl arachidate, eicosanoic acid, methyl ester, arachidic acid methyl ester, methyl eicosanoate, methyl arachisate, arachidic acid methylester, eicosanoic acid methyl ester, methylarachidate, methyl aracidate
methyl icosanoate

Specifications

1120-28-1
White
>110°C (230°F)
MFCD00009014
methyl arachidate, eicosanoic acid, methyl ester, arachidic acid methyl ester, methyl eicosanoate, methyl arachisate, arachidic acid methylester, eicosanoic acid methyl ester, methylarachidate, methyl aracidate
QGBRLVONZXHAKJ-UHFFFAOYSA-N
methyl icosanoate
14259
≥98%
Methyl eicosanoate
48°C
215°C (10 mmHg)
C21H42O2
2 g
Soluble in hot alcohol. Sparingly Soluble in water.
CCCCCCCCCCCCCCCCCCCC(=O)OC
326.57
326.56
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 214-304-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only