missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl levulinate, 99+%, Thermo Scientific Chemicals
$98.00 - $300.49
Chemical Identifiers
CAS | 624-45-3 |
---|---|
Molecular Formula | C6H10O3 |
Molecular Weight (g/mol) | 130.143 |
MDL Number | MFCD00017499 |
InChI Key | UAGJVSRUFNSIHR-UHFFFAOYSA-N |
Synonym | methyl levulinate, methyllevulinate, levulinic acid methyl ester, pentanoic acid, 4-oxo-, methyl ester, methyl 4-oxovalerate, levulinic acid, methyl ester, 4-oxovaleric acid methyl ester, 4-oxo-pentanoic acid methyl ester, methyl levulate, me ester |
PubChem CID | 69354 |
IUPAC Name | methyl 4-oxopentanoate |
SMILES | CC(=O)CCC(=O)OC |
Description
Methyl levulinate is mainly used as raw material on perfume.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
624-45-3 | |
130.143 | |
UAGJVSRUFNSIHR-UHFFFAOYSA-N | |
69354 | |
CC(=O)CCC(=O)OC |
C6H10O3 | |
MFCD00017499 | |
methyl levulinate, methyllevulinate, levulinic acid methyl ester, pentanoic acid, 4-oxo-, methyl ester, methyl 4-oxovalerate, levulinic acid, methyl ester, 4-oxovaleric acid methyl ester, 4-oxo-pentanoic acid methyl ester, methyl levulate, me ester | |
methyl 4-oxopentanoate |
Specifications
624-45-3 | |
191°C | |
Pleasant | |
MFCD00017499 | |
methyl levulinate, methyllevulinate, levulinic acid methyl ester, pentanoic acid, 4-oxo-, methyl ester, methyl 4-oxovalerate, levulinic acid, methyl ester, 4-oxovaleric acid methyl ester, 4-oxo-pentanoic acid methyl ester, methyl levulate, me ester | |
UAGJVSRUFNSIHR-UHFFFAOYSA-N | |
methyl 4-oxopentanoate | |
69354 | |
≥99% | |
Methyl levulinate |
1.05 g/mL | |
72°C (161.6°F) | |
C6H10O3 | |
25 g | |
It is soluble in ethanol,ether,acetone and benzene. | |
CC(=O)CCC(=O)OC | |
130.143 | |
130.14 | |
Liquid |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 210-846-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only