missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl linolenate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 368100050

 View more versions of this product

Catalog No. AC368100050

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methyl linolenate
301-00-8
0.8950g/mL
>110°C
98.5% min. (GC)
C19H32O2
CH3(CH2CH=CH)3(CH2)7CO2CH3
5 g
0.895
Solubility in water: insoluble
CC\C=C/C\C=C/C\C=C/CCCCCCCC(=O)OC
292.46
292.46
Liquid
99%
Colorless
182.0°C (3.0 mmHg)
Authentic
Glass bottle
1.4700 to 1.4720
MFCD00135851
02, IV, 1782
methyl linolenate, linolenic acid methyl ester, methyl alpha-linolenate, linolenic acid, methyl ester, unii-0s1ns923k6, alpha-linolenic acid methyl ester, methyl all-cis-9,12,15-octadecatrienoate, methyl 9z,12z,15z-octadeca-9,12,15-trienoate, 9,12,15-octadecatrienoic acid, methyl ester, z,z,z, methyl cis,cis,cis-octadec-9,12,15-trienoate
DVWSXZIHSUZZKJ-YSTUJMKBSA-N
methyl (9Z,12Z,15Z)-octadeca-9,12,15-trienoate
5319706
99%

Chemical Identifiers

301-00-8
292.46
DVWSXZIHSUZZKJ-YSTUJMKBSA-N
5319706
CC\C=C/C\C=C/C\C=C/CCCCCCCC(=O)OC
C19H32O2
MFCD00135851
methyl linolenate, linolenic acid methyl ester, methyl alpha-linolenate, linolenic acid, methyl ester, unii-0s1ns923k6, alpha-linolenic acid methyl ester, methyl all-cis-9,12,15-octadecatrienoate, methyl 9z,12z,15z-octadeca-9,12,15-trienoate, 9,12,15-octadecatrienoic acid, methyl ester, z,z,z, methyl cis,cis,cis-octadec-9,12,15-trienoate
methyl (9Z,12Z,15Z)-octadeca-9,12,15-trienoate
Safety and Handling

Safety and Handling

EINECSNumber : 206-102-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only