missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl linolenate, 99%, Thermo Scientific Chemicals

$163.90 - $711.97

Chemical Identifiers

CAS 301-00-8
Molecular Formula C19H32O2
Molecular Weight (g/mol) 292.46
MDL Number MFCD00135851
InChI Key DVWSXZIHSUZZKJ-YSTUJMKBSA-N
Synonym methyl linolenate, linolenic acid methyl ester, methyl alpha-linolenate, linolenic acid, methyl ester, unii-0s1ns923k6, alpha-linolenic acid methyl ester, methyl all-cis-9,12,15-octadecatrienoate, methyl 9z,12z,15z-octadeca-9,12,15-trienoate, 9,12,15-octadecatrienoic acid, methyl ester, z,z,z, methyl cis,cis,cis-octadec-9,12,15-trienoate
PubChem CID 5319706
IUPAC Name methyl (9Z,12Z,15Z)-octadeca-9,12,15-trienoate
SMILES CC\C=C/C\C=C/C\C=C/CCCCCCCC(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC368100010
SDS View Documents
Thermo Scientific Chemicals
368100010
1 g Glass bottle
Each for $163.90
 
AC368100050
SDS View Documents
Thermo Scientific Chemicals
368100050
5 g Glass bottle
Each for $711.97
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

301-00-8
292.46
DVWSXZIHSUZZKJ-YSTUJMKBSA-N
5319706
CC\C=C/C\C=C/C\C=C/CCCCCCCC(=O)OC
C19H32O2
MFCD00135851
methyl linolenate, linolenic acid methyl ester, methyl alpha-linolenate, linolenic acid, methyl ester, unii-0s1ns923k6, alpha-linolenic acid methyl ester, methyl all-cis-9,12,15-octadecatrienoate, methyl 9z,12z,15z-octadeca-9,12,15-trienoate, 9,12,15-octadecatrienoic acid, methyl ester, z,z,z, methyl cis,cis,cis-octadec-9,12,15-trienoate
methyl (9Z,12Z,15Z)-octadeca-9,12,15-trienoate

Specifications

301-00-8
0.8950g/mL
>110°C
98.5% min. (GC)
C19H32O2
CH3(CH2CH=CH)3(CH2)7CO2CH3
1 g
0.895
Solubility in water: insoluble
CC\C=C/C\C=C/C\C=C/CCCCCCCC(=O)OC
292.46
292.46
Liquid
Colorless
182.0°C (3.0 mmHg)
Authentic
Glass bottle
1.4700 to 1.4720
MFCD00135851
02, IV, 1782
methyl linolenate, linolenic acid methyl ester, methyl alpha-linolenate, linolenic acid, methyl ester, unii-0s1ns923k6, alpha-linolenic acid methyl ester, methyl all-cis-9,12,15-octadecatrienoate, methyl 9z,12z,15z-octadeca-9,12,15-trienoate, 9,12,15-octadecatrienoic acid, methyl ester, z,z,z, methyl cis,cis,cis-octadec-9,12,15-trienoate
DVWSXZIHSUZZKJ-YSTUJMKBSA-N
methyl (9Z,12Z,15Z)-octadeca-9,12,15-trienoate
5319706
99%
Methyl linolenate, 99%
Safety and Handling

Safety and Handling

EINECSNumber : 206-102-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only